Learn More
Abnova™ USP6 (Texas Red/FITC) Split FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
$2394.00
Specifications
Source | Genomic DNA |
---|---|
Location | 17p13.2/17p13.2 |
Description | Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique. |
Includes | DAPI Counterstain (1500 ng/mL ) 250 uL |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-027-352
|
Abnova™
FS0028 |
200 μL |
Each of 1 for $2,394.00
|
|
|||||
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Genomic DNA | |
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique. | |
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections) | |
Approx. 530kb/590kb |
17p13.2/17p13.2 | |
DAPI Counterstain (1500 ng/mL ) 250 uL | |
USP6 Split FISH Probe |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.