Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP9X Polyclonal Antibody, CoraLite594, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc CL59455054100UL

Description
USP9X Polyclonal antibody specifically detects USP9X in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
USP9X | |
Polyclonal | |
CoraLite 594 | |
Q93008 | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunocytochemistry | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
5730589N07Rik; AA407302; AA407699; AL022658; AL022749; DFFRX; FAF; FAF-X; Fafl; FAM; MRX99; MRXS99F; RGD1560056; USP9; USP9X | |
Antigen affinity chromatography | |
RUO | |
8239 | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction