Learn More
ATHENS RESEARCH AND TECHNOLOGY INC Yamato 27L Vacuum Oven, Benchtop, 220V
Supplier:  ATHENS RESEARCH AND TECHNOLOGY INC ADP310C
Encompass_Preferred
Standard small size 27L benchtop vacuum drying oven with enhanced safety features. Easy input of parameters and settings. Digital PID controller supports fixed temperature, quick auto-stop, auto stop, auto start and program operations. Self-diagnostic and overheating prevention functions. Silicon rubber door seal prevents air from leaking. Independent over heating prevention device for each circuit. Customizable with N₂ gas inlet and communication ports. Calibration off-set function. Easy maintenance. Operating temp. range of 40~240°C
Catalog No. 50-189-8032
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.