Learn More
GELITA USA VacciPro, 1 kg
Supplier:  GELITA USA VACCIPRO
Encompass
VacciPro’s low allergenic potential, high tolerance by the body and an affinity for tissue cells, it is an ideal substance for injected pharmaceuticals. With high stabilization properties, VacciPro is recognized by a number of global vaccine producers as the gold standard in vaccine development.
Catalog No. NC0834969
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.