Learn More
Foxx Life Sciences Vactrap™ Vacuum Trap Bottle (Vacuum Flask Alternative) for Aspiration and Vacuum Protection, Shatterproof 4 Liter Aspirator Bottle Assembly, Autoclavable Polypropylene (PP), 4L Bottle with Inline Vent Filter, No Secondary Container
SDP
Supplier:  Foxx Life Sciences 3054002FLS
Encompass_Preferred
305-4002-FLS: Vactrap™, PP, 4L, No Bin, ID TubingFoxx Life Sciences Vactrap is the worlds most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap comes ready to usesimply attach to your vacuum and begin your work. The full Vactrap system includes two Foxx Heavy Duty Vacuum Bottles (autoclavable PP or chemically-resistant HDPE), each with a patented VersaCap closure for tangle-free openingno need to disconnect tubing, ever. A 0.2m PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nested neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for modulation of vacuum. Other Vactrap options offer one bottle, or setups without secondary containment. Say goodbye to glass. Say hello to Vactrap.
Catalog No. 50-148-9328
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.