Learn More
Foxx Life Sciences Vactrap XL™ Secondary Container Spill Basin, Safety Tray with Dividers, for use with VactrapXL™ Vacuum Trap System or for General Lab Safety, Waste Storage, and Spill Containment
Supplier: Foxx Life Sciences 3000006FLS
VactrapXL™ Red Bin w/ DividersFoxx Life Sciences Vactrap is the world’s most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap comes ready to use—simply attach to your vacuum and begin your work. The Vactrap Bin serves as a super-stable basin for secondary containment of the Vactrap system. Complete with dividers to hold bottles firmly in place, the bin vastly increases stability while maintaining a small footprint. Also great for use for general secondary containment outside of vacuum or aspiration applications.Say goodbye to glass. Say hello to Vactrap.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.