Learn More
Electron Microscopy Sciences Vacuum Oven Model 3618-5, 120 V, 14"W x 14"H x 20"D chamber, 2.3 cu. ft (65 L) Capacity
Supplier:  Electron Microscopy Sciences 6323810
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Multipurpose vacuum oven with 220 deree C maximum temperature conserves chamber space for drying, curing, vacuum embedding, and plating applications. Silicone door gasket and a positive latch door maintains the seal for all vacuum levels. Exclusive Polycarbonate safety shield protects door glass. Two removable aluminum shelves including standard. Easy-to-clean #304 staniless steel chamber interior. Powder coated heavy guage steel exterior construction.
Chamber: 14"W x 14"H x 20"D
Capacity: 2.3 cu. ft (65 L)
Thermometer: LED
Catalog No. 50-313-08
missing translation for 'QuantityLeft'