Learn More
Chemglass Life Sciences Vacuum Trap, Complete, Rodaviss Threaded Joint
SDP
Supplier:  Chemglass Life Sciences CG453201
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Vacuum Trap, Complete, Rodaviss Threaded Joint. Vacuum trap with a 45/50 Rodaviss joint. Threaded joint provides a vacuum tight seal without grease and if frozen, can be taken apart using the optional loosening ring. Serrated hose connections are 13.5mm at the smallest serration and 15.5mm at the largest serration. Complete trap is supplied with stopper, lower body, viton o-ring, loosening ring, and rodaviss cap.
Catalog No. 50-873-462