Learn More
ValueMax™ Easy-Breathe™ Lab Jackets, Ceil Blue
Click to view available options
Size:
3X-Large
4X-Large
Description
- Silky soft 'SMS' fabrics
- Anti-static treated and no clinging
- Snap front with three pockets, knit cuff and collar
- Hip length and bright colors for any operations
- Meet AAMI level 2 fluid resistant standard
Specifications
Specifications
Size | 3X-Large |
Gender | Unisex |
Color | Ceil Blue |
Collar Type | Knit |
Cuff Style | Knit |