Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Ceil Blue
Catalog No. p-3148644
Click to view available options
Size:
Large
X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Static free — no clinging
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Size 5X-Large
Material Polypropylene
Color Ceil Blue
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.