Learn More
Chemglass Life Sciences Valve, Chem-Vac™, Hi-Vac, 180°, 0-12mm, Viton O-Ring
SDP
Supplier:  Chemglass Life Sciences CG92703
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Valve, Chem-Vac™, Hi-Vac, 180°, 0-12mm, Viton O-Ring. CHEM-VAC™ valve with two sidearms sealed to the barrel at a 180º angle to one another and on the same plane. Valve has TFE plug with three o-rings, the tip o-ring making the primary seal on the precision formed tapered seat. O-rings on the non-rotating shaft form an excellent dynamic seal between the TFE plug and glass barrel. See CG-935 for replacement plug, CG-537 for o-ring replacement kit, and CG-904 for glass portion only.
Catalog No. CG92703
missing translation for 'QuantityLeft'