Learn More
TARGETMOL CHEMICALS INC VANOXERINE DIHYDROCHLORI 50MG
Supplier:  TARGETMOL CHEMICALS INC T7153
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vanoxerine dihydrochloride (GBR-12909 dihydrochloride) is a potent inhibitor that blocks dopamine uptake (IC50 1-51 nM)purity: 97%
Catalog No. 50-257-6344
missing translation for 'QuantityLeft'