Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Jars and Pails
Collect effluent from multiple sources while minimizing harmful solvent exhaust
Supplier:  Cole-Parmer™ 1202098
 View more versions of this product
Catalog No. 13-245-161
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Recover clean extracts and shorten the time spent on method development using TELOS neo polymeric SPE columns.
  • High surface area and capacity allow the use of smaller sorbent masses, leading to reduced elution volumes and evaporation/reconstitution steps.
  • Polymeric sorbents are water-wettable and not affected by drying out, providing better flow characteristics, retention, and elution.
  • Columns provide high and reproducible recoveries for a wide range of analytes, including acidic, neutral, basic, and multi-charged analytes.
Specifications
Specifications
Solvent Waste Kit
Polypropylene
70 mm Rieke 6TPI
1 x 0.5 in. NPT(F), 3 x 0.25 in. NPT(F), and 6 x 1/4-28 UNF(F)
Product Suggestions
Product Suggestions