Learn More
Walter Products™ Variable Volume 8 Channel Micropipettes
SCI_ED
Catalog No. p-7225987 Shop All Walter Products Products
Click to view available options
Accuracy:
For Use With (Equipment):
Increments:
Volume (Metric):
These multichannel micropipettes are lightweight and convenient to operate, requiring little pipetting strength.
  • High chemical and UV resistance
  • Completely autoclavable at 121°C
  • 4-position volume settings and a volume display which can easily be read
  • True one-hand operation for both right and left handed operators
  • Rotating dispensing head for optimal pipetting convenience
  • Due to optimized design on the tip cone, compatible with a wide range of tips
Specifications
Accuracy
±0.8/1.4/6%
Autoclavable
Autoclavable
For Use With
200 μL Tip
Number of Channels
8
Type
Micropipette
Volume (Metric)
5 to 50 μL
For Use With (Equipment)
200 μL Tips
Increments
0.05 μL