Learn More
TARGETMOL CHEMICALS INC VATANIDIPINE 5MG
Supplier:  TARGETMOL CHEMICALS INC T24932
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vatanidipine (AE0047) is a novel dihydropyridine (DHP)-type calcium channel blocker with slow-onset pharmacological actions.A slow-onset and long-lasting hypotensive action was observed in various experimental hypertensive models. purity: 100%
Catalog No. 50-258-0776
missing translation for 'QuantityLeft'