Learn More
OBORO LABS INC VEIGE CATALYST 1
Supplier:  OBORO LABS INC OBOCATV1.10G
Encompass_Preferred
The trianionic pincer tungsten-alkylidyne Veige Catalyst is used in the synthesis of cyclic polymers. The Veige Catalyst will polymerize alkynes to produce cyclic polyacetylenes. The metal carbon bond coupled with a strained trianionic pincer ligand creates a highly reactive complex that will react with most unsaturated substrates. The ligands are specifically designed to provide a unique coordination environment, influencing the molecule's properties and potential applications. The presence of tetrahydrofuran ligands further enhances its solubility and compatibility in organic solvents, expanding its applicability in diverse chemical processes. ApplicationsThe distinctive structure of the Veige Catalyst makes it a promising candidate for various catalytic applications.
Catalog No. NC2817217
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.