Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals (±)-Verapamil hydrochloride, 99+%

Catalog No. AC329330050
Click to view available options
Quantity:
Packaging:
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

152-11-4

Molecular Formula

C27H38N2O4·HCl

Molecular Weight (g/mol)

491.07

MDL Number

MFCD00055208

InChI Key

DOQPXTMNIUCOSY-UHFFFAOYSA-N

Synonym

verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril

PubChem CID

62969

IUPAC Name

2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride

SMILES

CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl

Specifications

CAS Min %

99.0

CAS Max %

100.0

Melting Point

140.0°C to 144.0°C

Color

White

pH

4.5 to 6.5 (5% soln.)

Ignition Residue

0.1% max.

Loss on Drying

0.5% max. (105°C, 2 hrs)

Infrared Spectrum

Authentic

Assay Percent Range

99+%

Packaging

Glass bottle

Quantity

5 g

Merck Index

15, 10144

Solubility Information

Solubility in water: soluble in water. Other solubilities: soluble in ethanol

Formula Weight

491.07

Percent Purity

99+%

Physical Form

Powder

Chemical Name or Material

(±)-Verapamil hydrochloride

Hazard Category

GHS Signal Word: Danger

Hazard Statement

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.

Precautionary Statement

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

EINECSNumber

205-800-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.