Learn More
PBL Assay Science VeriKine-HS Human Interferon Beta (IFN-Beta, IFNb) ELISA, Tissue Culture Media (TCM) (High Sensitivity) [1x96-well plate]
Supplier:  PBL Assay Science 414351
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
VeriKine-HS Human Interferon Beta ELISA Kit, TCM is manufactured by PBL Assay Science (Catalog #41435-1) for research use only (RUO) and packaged as a 1x96-well plate. This IFNb ELISA is Tissue Culture Media (TCM) compatible, quantifies Hu IFN-Beta, and has an assay range of 2.34-150 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.
Catalog No. NC1813836
missing translation for 'QuantityLeft'