Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Copper (Cu) Standard for AAS, 1000ppm in 2% HNO3, ISO 17025/Guide 34, Ricca Chemical
SDP

Catalog No. p-7073903 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100mL
500mL

Formulated for high performance and reliable results

  • 1000ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 3251-23-8, 7697-37-2, 7732-18-5
Molecular Formula CuN2O6
Molecular Weight (g/mol) 187.554
InChI Key XTVVROIMIGLXTD-UHFFFAOYSA-N
PubChem CID 18616
ChEBI CHEBI:78036
IUPAC Name copper;dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]

Specifications

CAS Min % 95.746%, 1.96%, 0.294%
CAS Max % 99.654%, 2.04%, 0.306%
Physical Form Liquid
Grade High Purity
Packaging Bottle, Poly Natural
Quantity 100mL
Specific Gravity 1.01
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.