Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ ICP Tuning Solution 15, 500ppm: K; 50ppm: Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn; ISO 17025/Guide 34, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
100mL
500mL
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
CAS | 7697-37-2, 13473-90-0, 13548-38-4, 3251-23-8 |
CAS Min % | 0.98%, 0.0686%, 0.0196%, 0.0098% |
CAS Max % | 1.02%, 0.0714%, 0.0204%, 0.0102% |
Physical Form | Liquid |
Grade | High Purity |
Packaging | Bottle, Poly Natural |
Quantity | 100mL |
Specific Gravity | 1.08 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction