Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ Interference Check Solution 12, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
100 mL
500 mL
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
Packaging | Natural LDPE Bottle |
CAS | 7697-37-2, 13473-90-0 |
Quantity | 100 mL |
CAS Min % | 4.9%, 2.7244% |
CAS Max % | 5.1%, 2.8356% |
UN Number | UN3264 |
Grade | High Purity |
Specific Gravity | 1.07 |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction