Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ Internal Calibration Standard 7, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
100 mL
500mL
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
Physical Form | Liquid |
CAS Min % | 4.9%, 0.009702% |
CAS Max % | 5.1%, 0.010098% |
CAS | 7790-69-4, 10035-06-0, 7697-37-2 |
UN Number | UN3264 |
Grade | High Purity |
Packaging | Natural LDPE Bottle |
Quantity | 100 mL |
Specific Gravity | 1.06 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction