Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Mercury (Hg) Standard for AAS, 1000ppm in 10% HNO3, ISO 17025/Guide 34, Ricca Chemical
SDP

Catalog No. p-7074013 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:

Formulated for high performance and reliable results

  • 1000ppm in 10% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS

10045-94-0, 7697-37-2, 7732-18-5

Molecular Formula

HgN2O6

Molecular Weight (g/mol)

324.60

MDL Number

MFCD00011038

InChI Key

ORMNPSYMZOGSSV-UHFFFAOYSA-N

PubChem CID

24864

IUPAC Name

mercury(2+) dinitrate

SMILES

[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging

Amber Glass Bottle

Quantity

100 mL

CAS Min %

88.0432%, 9.8%, 0.1568%

CAS Max %

91.6368%, 10.2%, 0.1632%

UN Number

UN2031

Grade

High Purity

Specific Gravity

1.02

Physical Form

Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.