Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Mercury Standard for ICP, 1000ppm in 10% HNO3, ISO 17025/Guide 34, Ricca Chemical
SDP

Catalog No. p-7074014 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100mL
250 mL
500 mL

Formulated for high performance and reliable results

  • 1000ppm in 10% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10045-94-0, 7697-37-2, 7732-18-5
Molecular Formula HgN2O6
Molecular Weight (g/mol) 324.6
InChI Key ORMNPSYMZOGSSV-UHFFFAOYSA-N
PubChem CID 24864
IUPAC Name mercury(2+);dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Hg+2]

Specifications

CAS Min % 88.0432%, 9.8%, 0.1568%
CAS Max % 91.6368%, 10.2%, 0.1632%
UN Number UN3264
Physical Form Liquid
Grade High Purity
Packaging Bottle, Amber Glass
Quantity 100mL
Specific Gravity 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.