Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ Mixed Cation Standard 6, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
50 mL
100mL
Description
- 50 ppm Li+; 200 ppm Na+, K+, Mg2+; 400 ppm NH4+, 1000 ppm Ca2+
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
Physical Form | Liquid |
CAS Min % | 0.98%, 0.392%, 0.1764%, 0.0784%, 0.0686%, 0.05% |
CAS Max % | 1.02%, 0.408%, 0.1836%, 0.0816%, 0.0714%, 0.05% |
CAS | 471-34-1, 6484-52-2, 7786-30-3, 7631-99-4, 7790-69-4, 7732-18-5 |
Grade | High Purity |
Packaging | Natural Poly Bottle |
Quantity | 50 mL |
Specific Gravity | 1.06 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction