Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Multi-Element Standard 10, 50ppm: As, K; 10ppm: La, Li, Mn, Ni, Sr, Zn; 1ppm Ba, Mg; ISO 17025/Guide 34, Ricca Chemical
SDP

Catalog No. p-7074026 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100mL
500mL

Formulated for high performance and reliable results

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

CAS Min % 1.96%, 0.0098%, 0.009702%, 0.00931%, 0.003234%, 0.00%
CAS Max % 2.04%, 0.0102%, 0.010098%, 0.00969%, 0.003366%, 0.00%
CAS 7757-79-1, 7790-69-4, 7778-39-4, 10377-66-9, 13138-45-9, 7697-37-2
Physical Form Liquid
Grade High Purity
Packaging Bottle, Poly Natural
Quantity 100mL
Specific Gravity 1.07
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.