Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ Multi-Element Standard 8; 50ppm K; 10ppm La, Li, Mn, Na, Sr; 1ppm Ba, Ca; ISO 17025/Guide 34, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
100 mL
250mL
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
Packaging | Natural LDPE Bottle |
Quantity | 100 mL |
CAS Min % | 1.96%, 0.0098%, 0.009702%, 0.003626%, 0.003234%, 0.00% |
CAS Max % | 2.04%, 0.0102%, 0.010098%, 0.003774%, 0.003366%, 0.00% |
CAS | 7790-69-4, 7631-99-4, 10377-66-9, 10277-43-7, 7757-79-1, 7697-37-2 |
Grade | High Purity |
Specific Gravity | 1.07 |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction