Learn More
Labelmaster Inc Vermiculite Grade 2A, 4-cu. ft. Bag
Supplier: Labelmaster Inc KTV400A2
Vermiculite is a great choice for providing absorbent cushioning when packaging hazardous materials that may accidentally leak or spill during shipping. Ideal for use with UN-certified wooden boxes and Variation Two shippers, this Grade 2-A Medium 4GV accessory is chemically inert, asbestos-free and can absorb liquids and spills of all kinds, approximately 1.75 cubic ft. of vermiculite absorbs three gallons of water, and absorption may be different for certain chemical spills. Sold as a 4-cubic-ft. bag.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.