Learn More
STERIS™ Vesta-Syde SQ 128 st RTU Disinfectant Wipes
Shop All STERIS Corporation ProductsVesta-Syde SQ 128 st Ready-to-Use Disinfectant Wipes combine knitted polyester wipes in a unique packaging system with a ready-to-use solution. The formula is contained in an internal pouch separate from the wipes to maximize chemical stability. This packaging system provides a simple, convenient and consistent method to saturate the wipes.
Vesta-Syde SQ 128 st Ready-to-Use Disinfectant Wipes kill a broad spectrum of common bacteria and fungi to meet your contamination control needs. Each package is double-bagged and gamma-irradiated for use in aseptic areas. Vesta-Syde SQ 128 st Ready-to-Use Disinfectant Wipes are part of the Vesta-Syde SQ family of products. These ready-to-use options offer maximum flexibility to simplify your validation activities.
WHY VESTA-SYDE READY TO USE DISINFECTANT WIPES?
- Reliability - Vesta-Syde SQ 128 st Ready-to-Use Disinfectant chemistry
- Ease of Use - Eliminates the need to validate dilution preparation procedures by providing saturated wipes with minimal operator effort
- Productivity - Saves time and labor for chemical preparation
- Efficacy - Microbial control against a broad spectrum of bacteria and yeast
- Convenience - Low linting wipes with sealed edges that are ideal for cleanroom applications. Gamma irradiated for easy introduction into aseptic areas
- Quality - Each lot is USP sterility tested and documented by a Certificate of Analysis
- Assurance - Manufactured in accordance with the cGMP with complete lot traceability
Specifications
Product Type
Disinfectant Wipe
Length (English)
9 in.
Length (Metric)
23 cm
Width (English)
9 in.
Width (Metric)
23 cm
Material
100% Knitted Polyester
For Use With (Application)
RTU Wipes
Packaging Quantity
180 Wipes/Cs.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.