Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VETRANAL™ Penicillin V potassium salt, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
:
250 mg
Description
Environmental and Pharmaceutical (small molecule)
Specifications
Specifications
Quantity | 250 mg |
CAS | 132-98-9 |
Type | Analytical Standard |
Product Line | VETRANAL |
Linear Formula | C16H17N2O5 SK |
MDL Number | MFCD00051771 |
Synonym | Phenoxymethylpenicillin potassium salt |
Formula Weight | 388.48 |
Grade | Analytical Standard |
Physical Form | Neat |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction