Learn More
TWD Scientific LLC BioX Tricode 0.5mL Cryovial, Purple Cap, External Thread. Case of 1000
SDP
Supplier:  TWD Scientific LLC BX889055
Encompass_Preferred
BioX 0.5mL clear polypropylene sterile pre-set 1D & 2D barcoded cryovials with external thread and purple caps assembled. Cryovials have writing patch and marked graduations. 25 cryovials/bag, 20 bags/pack, 2 packs/case, price/case
Catalog No. 50-227-4238
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.