Learn More
Chemglass Life Sciences Vial, Drosophila, 28.5 x 95mm, Wide
SDP
Supplier:  Chemglass Life Sciences CGE4789003
Encompass_Preferred
• Made from premium grade, transparent polystyrene• Narrow, wide drosophila vials are glass-like clarity for sample observation• Disposable vials are a safe and cost-effective alternative to glass vials• Narrow vials 25x95mm or wide vials 28.5x95mm available• Compatible with CGE-4790 Plugs (Sold Separately)
Catalog No. 50-189-7635
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.