Learn More
Vidaro™ B-Fore™ ESD Hoods
Catalog No. p-21756 Shop All Vidaro Corporation Products
Click to view available options
Color:
Light Blue
Navy
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Requires
Two female snaps on each cheek allow the attachment of a facial (not included).
High-performance fabric ideal for critical environments.
  • Super-dense weave of 100% microdenier polyester fibers minimizes particulate contamination
  • Microengineered carbon fiber grid dissipates electrostatic charges
  • Open-faced hood fits snugly under the chin, leaving the entire face exposed
  • Full-bib apron is long, to fit securely into a coverall or frock without pulling out.
  • Front closure has two stainless-steel snaps, and there are adjustment snaps at the back of the neck
Specifications
Product Type Open Face
Size X-Small
Material Polyester, Carbon
Color Light Blue
Closure Type Snap Front
Disposable Reusable
Model H3358
Quantity 1 Ea.
Includes Two snaps on each cheek, Stainless Steel front-snap closure, snap at back of neck for adjustment
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.