Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Vinegar
SCI_ED

Catalog No. p-4416665 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
3.8 L
Packaging:
Liquid - Cider
Liquid - White

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 64-19-7, 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless
Assay Percent Range 5% v/v
Formula Weight 60.05
Concentration or Composition (by Analyte or Components) 4 to 8% Acetic Acid Solution
Quantity 500 mL
Packaging Liquid - White
Physical Form Liquid
Chemical Name or Material Vinegar
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.