Learn More
Fisherbrand™ Vinyl Chair - Medium Bench Height with Medium Back, Seat Tilt
SDPSCI_ED
Fisherbrand™ Vinyl Chairs with Medium Back are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, larger back, arms, chrome adjustable foot ring, and self-braking casters. Grade A vinyl, in a variety of colors, offer additional antibacterial properties and a high-end look and feel.
 View more versions of this product
Catalog No. S0101377
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Medium Back
  • Adjustable from 21 to 28.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade A Vinyl
  • Additional antibacterial protections
  • High end look and feel
  • Variety of color options to suit any work space
Specifications
Specifications
General Purpose Seating
21 to 28.5 in.
Nylon Reinforced Fiberglass
Medium Bench Height
6-way adjustable up and down, in and out, twist toward or away from the user
25 in.
15 year parts, lifetime on pneumatic lift, 5 year on upholstery
Taupe
Yes
Round tube chrome foot ring with internal lock
19 in.
Yes
Yes
Yes (Pneumatic)
46 lb.
2.5 in.
300 lb.
20 in.
18 in.
Yes
Yes
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
Black dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Vinyl
Product Suggestions
Product Suggestions