Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin A palmitate, MP Biomedicals™

Catalog No. ICN10326325 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

79-81-2

Molecular Formula

C36H60O2

Molecular Weight (g/mol)

524.874

InChI Key

VYGQUTWHTHXGQB-FFHKNEKCSA-N

Synonym

retinyl palmitate, vitamin a palmitate, retinol palmitate, retinol, hexadecanoate, arovit, all-trans-retinyl palmitate, optovit-a, retinyl hexadecanoate, aquapalm, dispatabs tabs

PubChem CID

5280531

ChEBI

CHEBI:17616

IUPAC Name

[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] hexadecanoate

SMILES

CCCCCCCCCCCCCCCC(=O)OCC=C(C)C=CC=C(C)C=CC1=C(CCCC1(C)C)C

Specifications

Color

Yellow to Orange

Quantity

25 g

Formula Weight

524.9

Physical Form

Powder

Chemical Name or Material

Vitamin A palmitate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.