Learn More
Glass Expansion Viton O-rings for PE Avio D-Torch (PKT 10)
Supplier:  Glass Expansion 708031576
Encompass_Preferred
Viton O-rings for PE Avio D-Torch (PKT 10). The O-Ring removal tool (50-100-1636) facilitates the removal and installation of O-rings used with Glass Expansion D-Torches. This tool is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3041
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.