Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VOA Calibration Mix #2, Restek™
SDP

Catalog No. 06729033 Shop All Restek Corporation Products
Click to view available options
Quantity:
1 mL
1mL/ampule contains 2000μg/mL each in P and T methanol
  • Benzene (71-43-2)
  • Carbon disulfide (75-15-0)
  • Ethylbenzene (100-41-4)
  • Toluene (108-88-3)
  • Vinyl acetate (108-05-4)
  • o-Xylene (95-47-6)
  • p-Xylene (106-42-3)

Specifications

Concentration 2000μg/mL each in P and T Methanol
Packaging Ampule
Quantity 1 mL
CAS 61-73-4, 108-88-3, 108-05-4, 95-47-6, 71-43-2, 75-15-0, 100-41-4
UN Number 1993
Formula Weight 32.04
Explosion Limit Upper limit: 36% Lower limit: 6%
Concentration or Composition (by Analyte or Components) Methanol: 98.6%p-xylene: 0.2%Toluene: 0.2%Vinyl Acetate: 0.2%o-xylene: 0.2%Benzene: 0.2%Carbon Disulfide: 0.2%Ethylbenzene: 0.2%
Ignition Point 464°C
Odor Mild
Vapor Density 1.1 (air = 1)
Analyte or Component Names Benzene Carbon disulfide Ethylbenzene Toluene Vinyl acetate o-Xylene p-Xylene
Melting Point -98°C
Flash Point 12°C
Specific Gravity 0.791 to 0.792g/cm3 at 20°C
Solubility Moderately soluble
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.