Learn More
Medchemexpress LLC HY-100007 250mg Medchemexpress, Vonoprazan CAS:881681-00-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100007250MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-100007 250mg Vonoprazan CAS:881681-00-1 Vonoprazan (TAK-438 free base) is an orally active potassium-competitive acid blocker which inhibits H+, K+-ATPase activity with an IC50 of 19 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7708
missing translation for 'QuantityLeft'