Learn More
Medchemexpress LLC HY-15295 100mg Medchemexpress, Vonoprazan (Fumarate) CAS:881681-01-2 Purity:>98%
Medchemexpress, HY-15295 100mg Vonoprazan (Fumarate) CAS:881681-01-2 Vonoprazan Fumarate (TAK-438), a proton pump inhibitor (PPI), is a potent and orally active potassium-competitive acid blocker (P-CAB), with antisecretory activity. Vonoprazan Fumarate inhibits H+,K+-ATPase activity in porcine gastric microsomes with an IC50 of 19 nM at pH 6.5. Vonoprazan Fumarate is developed for the research of acid-related diseases, such as gastroesophageal reflux disease and peptic ulcer disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.