Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VU 0357017 hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Positive allosteric modulator of muscarinic M1 receptors (EC50 = 198 nM). Displays no activity at M2-M5 at concentrations up to 30 μM. Potentiates NMDA receptor currents in hippocampal neurons; activity reverses cognitive decifits in a rodent model of hippocampal-dependent memory. CNS penetrant.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 369.89 |
Percent Purity | >98% |
Chemical Name or Material | VU 0357017 hydrochloride |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction