Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VU 0364739 hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective phospholipase D2 (PLD2) inhibitor (IC50 = 20 nM); displays 75-fold selectivity against PLD1 (IC50 = 1500 nM).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 482.98 |
Percent Purity | >98% |
Chemical Name or Material | VU 0364739 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction