Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VU 10010, Tocris Bioscience™

Catalog No. p-4784780 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective allosteric potentiator of M4 receptors

Selective allosteric potentiator of M4 acetylcholine receptors (EC50 values are 33 and 0.7 nM for ACh in the absence and presence of VU10010 respectively). Binds to an allosteric site on the receptor increasing affinity for ACh and coupling to G-proteins.

Chemical Identifiers

CAS 633283-39-3
Molecular Formula C17H16ClN3OS
Molecular Weight (g/mol) 345.845
InChI Key FPRULFHDSFKYBV-UHFFFAOYSA-N
Synonym 3-amino-n-4-chlorophenyl methyl-4,6-dimethylthieno 2,3-b pyridine-2-carboxamide, 3-amino-n-4-chlorobenzyl-4,6-dimethylthieno 2,3-b pyridine-2-carboxamide, 2fli, 2-fly, f-ligrlo-nh2, 2-fap, 2-f-ligrlo-nh2, d0n2vn, 2-furoyl-ligrlo-amide trifluoroacetate salt, 2-furoyl-leu-ile-gly-arg-leu-orn-nh2 trifluoroacetate salt
PubChem CID 714286
IUPAC Name 3-amino-N-[(4-chlorophenyl)methyl]-4,6-dimethylthieno[2,3-b]pyridine-2-carboxamide
SMILES CC1=CC(=NC2=C1C(=C(S2)C(=O)NCC3=CC=C(C=C3)Cl)N)C

Specifications

Quantity 10 mg
Formula Weight 345.85
Percent Purity >99%
Chemical Name or Material VU 10010
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.