Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VU 152100, Tocris Bioscience™

Catalog No. p-4784782 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Positive allosteric modulator of M4 receptors

Selective positive allosteric modulator of M4 muscarinic acetylcholine receptors (mAChRs) (EC50 = 380 nM). Induces 21-fold shift in ACh potency at M4 receptor. Displays no activity at other mAChR subtypes.

Chemical Identifiers

CAS 409351-28-6
Molecular Formula C18H19N3O2S
Molecular Weight (g/mol) 341.429
InChI Key MDNWGCQSCGNTKH-UHFFFAOYSA-N
Synonym 3-amino-n-4-methoxyphenyl methyl-4,6-dimethylthieno 2,3-b pyridine-2-carboxamide, 3-amino-n-4-methoxybenzyl-4,6-dimethylthieno 2,3-b pyridine carboxamide, kinome_1269, d0k3hy, vu hplc, 3-amino-4,6-dimethyl-n-p-anisyl-thieno 2,3-b pyridine-2-carboxamide, 3-amino-4,6-dimethylthiopheno 2,3-b pyridin-2-yl-n-4-methoxyphenyl methyl carboxamide, 3-amino-n-4-methoxyphenyl methyl-4,6-dimethyl-2-thieno 2,3-b pyridinecarboxamide, 3-amino-n-4-methoxyphenyl methyl-4,6-dimethyl-thieno 2,3-b pyridine-2-carboxamide, 3-amino-n-4-methoxyphenyl methyl-4,6-dimethylthieno 5,4-b pyridine-2-carboxamide
PubChem CID 864492
IUPAC Name 3-amino-N-[(4-methoxyphenyl)methyl]-4,6-dimethylthieno[2,3-b]pyridine-2-carboxamide
SMILES CC1=CC(=NC2=C1C(=C(S2)C(=O)NCC3=CC=C(C=C3)OC)N)C

Specifications

Quantity 10 mg
Formula Weight 341.43
Percent Purity >98%
Chemical Name or Material VU 152100
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.