Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

W-7 hydrochloride, Tocris Bioscience™
GSA_VA

Catalog No. 0369100 Shop All R&D Systems Products
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Calmodulin antagonist. Inhibits myosin light chain kinase

W-7 hydrochloride is a calmodulin antagonist. Inhibits Ca2+-calmodulin-dependent phosphodiesterase (IC50 = 28 μM) and myosin light chain kinase (IC50 = 51 μM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 61714-27-0
Molecular Formula C16H22Cl2N2O2S
Molecular Weight (g/mol) 377.32
MDL Number MFCD00012559
InChI Key OMMOSRLIFSCDBL-UHFFFAOYSA-N
Synonym w-7 hydrochloride, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide hydrochloride, w-7, hydrochloride, n-6-aminohexyl-5-chloronaphthalene-1-sulfonamide hydrochloride, c16h21cln2o2s hydrochloride, 1-naphthalenesulfonamide, n-6-aminohexyl-5-chloro-, monohydrochloride, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide monohydrochloride, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide hcl, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide, hcl, n-aminohexyl-5-chloro-1-napthalenesulfonamide hydrochloride
PubChem CID 124887
IUPAC Name hydrogen N-(6-aminohexyl)-5-chloronaphthalene-1-sulfonamide chloride
SMILES [H+].[Cl-].NCCCCCCNS(=O)(=O)C1=C2C=CC=C(Cl)C2=CC=C1

Specifications

Quantity 100 mg
Formula Weight 377.33
Percent Purity >98%
Chemical Name or Material W-7 hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.