Learn More
Uline 48 x 18 x 34"" | Shelf Capacity: 250lbs |Open wire design reduces dust buildup.Wire shelves adjust in 1"" increments.NSF certified.Includes wall mounting hardware.Chrome finish resists rust and corrosion. Chrome-plated carbon steel
Supplier:  Uline H3026
Encompass
48 x 18 x 34" | Shelf Capacity: 250lbs |Open wire design reduces dust buildup.Wire shelves adjust in 1" increments.NSF certified.Includes wall mounting hardware.Chrome finish resists rust and corrosion. Chrome-plated carbon steel
Catalog No. NC2201605
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.