Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water with 0.1% (v/v) Formic Acid, Hypergrade, For LC-MS LiChrosolv™, MilliporeSigma™

Catalog No. p-7221264 Shop All MilliporeSigma Products
Click to view available options
Quantity:
2.5 L
4 L

Specifications

Boiling Point 100°C (1013 hPa)
Grade Hypergrade
Packaging Glass Bottle
Quantity 2.5 L
Recommended Storage Store at 2 to 30 °C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.