Learn More
Quality Biological Inc Water, Cell Culture Grade, 10 Liters
SDP
Supplier:  Quality Biological Inc 118162151
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
QBI¿s Cell Culture Grade Water is high quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. ¿ Endotoxin < 0.03 EU/mL ¿ Mycoplasma Tested ¿ Also used to dilute buffers and suspend cells. ¿ Prepared by reverse osmosis ¿ Passed through fine carbon ¿ Deionized through two resin beds ¿ Serially filtered twice through 0.1 ¿m positively charged membranes ¿ Endotoxin level < 0.03 EU/mL. ¿ Storage Condition: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Endotoxin and Mycoplasma Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 41116113 ¿ ¿ Max Shelf Life: 24 months
Catalog No. 50-751-7170