Learn More
Quality Biological Inc Water, Molecular Biology, CS 10 X 500 mL
SDP
Supplier:  Quality Biological Inc 351029101CS
Encompass_Preferred
QBI's Molecular Biology Water is prepared by reverse osmosis • Passed through fine carbon • Deionized through two resin beds • Serially filtered through 0.1 µm positively charged membranes • Endotoxin level less than 0.03 EU/mL. • Applications: Useful in reconstituting nucleic acids and/or proteins, RNA applications (i.e. Preparation of an RNA probe), DNA applications (i.e. Subcloning in pUC plasmids), and protein applications (i.e. Western Blotting). • Also, useful as a diluent or solvent. • Storage Condition: 15°C to 30°C • Shipping Condition: ambient temperature • Sterile Filtered & Deionized • Endotoxin (<0.03 EU/mL) and Mycoplasma Tested • DNase, RNase, and Protease Tested • Product and/or packaging contains recycled/renewable content • Product and/or packaging is recyclable or reduces waste • Product and/or packaging is safer to human and environmental health • UNSPSC: 41116134 • Harmonization Code: 3822.00.5090 • ECCN: EAR99 • Max Shelf Life: 36 months
Catalog No. 50-751-7260
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.