Learn More
*PMW INC B. Braun #R5000-01 Irrigation Solution Sterile Water for Irrigation Not for Injection Bottle 1,000 ML WATER STR, IRR SOL 1000ML (16/CS)
Supplier: *PMW INC 00264210100
Distilled (still contains some ions) PIC™ is a PVC-free, DEHP-free Plastic Irrigation Container, Bottle is designed for easy non-drip pouring, The closure is sterile with recessed set port, Bottle has a molded hand grip for easier container handling,
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.